Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin-1 Monoclonal Antibody (2E12D10), Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 679921IG150UL

Description
Talin-1 Monoclonal antibody specifically detects Talin-1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Talin-1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ILWEQ; KIAA1027; TLN; TLN1 | |
Protein G | |
RUO | |
7094 | |
-20°C | |
Liquid |
Western Blot | |
2E12D10 | |
Unconjugated | |
Q9Y490 | |
Mouse | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction