Learn More
Bevco Precision Manufacturing Co Everlast Tall Height ESD/ISO 4 Cleanroom Black Polyurethane Chair, Articulating Seat & Back Tilt; Polished Aluminum Base w/Adjustable Footring
Supplier: Bevco Precision Manufacturing Co 7551E1
BEVCO's Everlast ESD/ISO 4 Cleanroom chairs are designed for outstanding comfort and long-lasting use in highly technical environments. The black ESD polyurethane seat and back are static dissipative and are impervious to water, oil, and most chemicals, puncture resistant and easy to clean. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. The waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 20. 5" - 30. 5". This chair comes with a sturdy 27" diameter polished aluminum base with brass drag chain and adjustable ESD footring. Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.