Learn More
Bevco Precision Manufacturing Co Integra Tall Height ISO 6 Cleanroom Blue Vinyl Chair; Medium Back; Non-Tilt; Polished Aluminum Base w/Adjustable Footring
BEVCO’s Integra ISO 6 Cleanroom Series is an upholstered chair that offers around-the-clock reliability for work environments which need to meet particulate goals of ISO 14644-1. The medium backrest is specifically shaped to provide ample back support and comes standard with a heavy-duty polypropylene back shell to protect the upholstery. The generous seat provides an exceptional seating experience while the thick, resilient, high-density foam provides long-lasting comfort. This non-tilt model features manual back and seat height adjustments for increased comfort during extended sitting durations. The pneumatic height adjustment allows you to easily change your seat height from 21. 5" - 31. 5"Blue vinyl meets CAL 117 standards. This chair comes with a sturdy 27" diameter polished aluminum base with adjustable chrome footring. Model comes standard with mushroom glides. Optional casters can be ordered separately.