Learn More
AdipoGen Tamoxifen citrate salt
SDP
Supplier:  AdipoGen CDXT0201G001
Encompass_Preferred
Chemical. CAS 54965-24-1. Formula C26H29NO . C6H8O7. MW 563.64. Synthetic. Selective estrogen receptor modulator SERM used as adjuvant therapy for estrogen-dependent breast cancer. Antagonist of ER action in breast tissue and breast cancer cells and ER agonist in bone, uterus, and the cardiovasculature system. This prodrug is metabolized by cytochrome P450 CYP450 enzymes to the active metabolites N-desmethyl-TMX, 4-hydroxy-N-desmethyl-TMX endoxifen and 4-hydroxy-TMX afimoxifene. N,N-Didesmethyl-4-hydroxytamoxifen norendoxifen, another active metabolite has been found to act as a potent competitive aromatase inhibitor and may also be involved in its antiestrogenic activity. Binds microsomal antiestrogen binding sites, altering cholesterol esterification at therapeutic doses and impacting breast cancer cell differentiation, apoptosis, and autophagy. Protein kinase C PKC inhibitor and anti-angiogenetic factor.
Catalog No. 50-255-9965
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.