Learn More
Lumiprobe TAMRA DBCO 5-isomer 5mg
Supplier: Lumiprobe 271F0
This product is a derivative of tetramethylrhodamine (TMR, TAMRA) containing a cyclooctyne moiety (dibenzocyclooctyne, DBCO). Pure 5-isomer.DBCO reacts quickly and efficiently with azides by simply mixing the components without the need for a copper catalyst (so-called sterically promoted cycloaddition reaction (SPAAC)).TAMRA DBCO can be used for the labeling of proteins, peptides, nucleic acids, and other molecules containing azide groups. TAMRA is often used as a FRET acceptor for FAM fluorophore. Can replace DyLight 549.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.