Learn More
MP Biomedicals™ Taq Core Kit 25
Taq DNA Polymerase + dNTPs in one kit
Supplier:  MP Biomedicals™ 11EPTQK255C
 View more versions of this product
Catalog No. MPEPTQK255C
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
  • MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
  • dNTPs at 25mM each
Specifications
Specifications
5 x 250 u
Product Suggestions
Product Suggestions