Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tartaric Acid, Granular, FCC, 99.7-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Tartaric Acid, Granular, FCC is used as an leavening additive and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. (CAS No. 87-69-4, Molecular Formula: C4H6O6) Tartaric Acid, Granular, FCC
Specifications
Specifications
CAS Max % | 1 |
Percent Purity | 99.7 to 100.5% |
Grade | FCC |
Quantity | 2.5 kg |
Ignition Residue | 0.0005 |
Loss on Drying | 0.005 |
Optical Rotation | +12.0° to +13.0° |
Packaging | Poly Pail |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction