Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC-E 5005, Tocris Bioscience™

Catalog No. p-4817129 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PDE10A inhibitor

Potent and selective PDE10A inhibitor (IC50 values are 7.28, 239, 779, 919, 3100 and 3700 nM at PDE10A, 2A, 11A, 5A, 7B and 3A respectively and >5000 nM at PDE1B, 4A, 6, 8A and 9A). Reverses MK 801-induced hyperactivity in vivo.

Chemical Identifiers

CAS 959705-64-7
Molecular Formula C15H18N4O
Molecular Weight (g/mol) 270.336
InChI Key YNADXFWEXJTQSZ-UHFFFAOYSA-N
Synonym 2-methoxy-6,7-dimethyl-9-propylimidazo 1,5-a pyrido 3,2-e pyrazine, 2-methoxy-6,7-dimethyl-9-propyl-imidazo 1,5-a pyrido 3,2-e pyrazine, 3,4-dimethyl-8-methoxy-1-propyl-imidazo 1,5-a pyrido 3,2-e pyrazine, imidazo 1,5-a pyrido 3,2-e pyrazine,2-methoxy-6,7-dimethyl-9-propyl, 12-methoxy-5,7-dimethyl-3-propyl-2,4,8,13-tetraazatricyclo 7.4.0.0 2 ,? trideca-1 13 ,3,5,7,9,11-hexaene
PubChem CID 24802717
SMILES CCCC1=NC(=C2N1C3=C(C=CC(=N3)OC)N=C2C)C

Specifications

Quantity 10 mg
Formula Weight 270.33
Percent Purity >99%
Chemical Name or Material TC-E 5005
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.