Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC-F 2, Tocris Bioscience™

Catalog No. p-4816556 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Potent, reversible and selective FAAH inhibitor

Potent, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50 values are 28 and 100 nM for human and rat FAAH respectively). Noncovalent inhibitor; displays selectivity for FAAH over cannabinoid-related targets (IC50 > 20 μM for CB1, CB2 and TRPV1). Active in vivo.

Chemical Identifiers

CAS 1304778-15-1
Molecular Formula C26H25N5O2
Molecular Weight (g/mol) 439.519
InChI Key NXTBLPPTZRPJCA-IBGZPJMESA-N
Synonym tc-f 2, 1-3s-1-4-2-benzofuranyl-2-pyrimidinyl-3-piperidinyl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-3s-1-4-1-benzofuran-2-yl pyrimidin-2-yl piperidin-3-yl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-ethyl-3-1-4-benzofuran-2-yl pyrimidine-2-yl piperidine-3beta-yl-1h-benzoimidazole-2 3h-one, qj9
PubChem CID 25198728
IUPAC Name 1-[(3S)-1-[4-(1-benzofuran-2-yl)pyrimidin-2-yl]piperidin-3-yl]-3-ethylbenzimidazol-2-one
SMILES CCN1C2=CC=CC=C2N(C1=O)C3CCCN(C3)C4=NC=CC(=N4)C5=CC6=CC=CC=C6O5

Specifications

Quantity 10 mg
Formula Weight 439.51
Percent Purity >99%
Chemical Name or Material TC-F 2
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.