Learn More
Gold Biotechnology Inc TCEP-HCl, 51805-45-9, MFCD00145469, 10 g
Supplier: Gold Biotechnology Inc 51805459
Molecular Formula: C9 H15 O6 P . Cl H, Molecular Weight: 286.646, Store desiccated at -20°C, Synonym: tris(2-Carboxyethyl)phosphine, HCl. TCEP-HCl is an odorless reducing agent suitable for protein applications and in the preparation for electrophoresis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.