Learn More
Gold Biotechnology Inc TCEP-HCl, 25 g
SDP
Supplier:  Gold Biotechnology Inc TCEP25
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
TCEP25TCEP (Tris (2-Carboxyethyl) phosphine Hydrochloride) is an odorless reducing agent suitable for protein applications and in the preparation for electrophoresis. TCEP is water soluble, is stable in a variety of solutions and is particularly useful in the labeling of cysteine residues with maleimides, preventing the cysteines from forming disulfide bonds. As a reducing agent, TCEP has many other desirable features.
Catalog No. 50-153-2844
missing translation for 'QuantityLeft'