Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Telenzepine dihydrochloride, Tocris Bioscience™

Catalog No. p-4785660 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective M1 antagonist

Selective high affinity muscarinic M1 receptor antagonist (Ki = 0.94 nM). Inhibits gastric acid secretion.

Chemical Identifiers

CAS 147416-96-4
Molecular Formula C19H24Cl2N4O2S
Molecular Weight (g/mol) 443.387
InChI Key JTQJFSQQHGPLOX-UHFFFAOYSA-N
Synonym telenzepine dihydrochloride, telenzepine hydrochloride, prestwick_649, 4,9-dihydro-3-methyl-4-4-methyl-1-piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 4,9-dihydro-3-methyl-4-4-methyl-1, piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 1-methyl-10-2-4-methylpiperazin-1-yl acetyl-5h-thieno 3,4-b 1,5 benzodiazepin-4-one dihydrochloride
PubChem CID 6419995
IUPAC Name 1-methyl-10-[2-(4-methylpiperazin-1-yl)acetyl]-5H-thieno[3,4-b][1,5]benzodiazepin-4-one;dihydrochloride
SMILES CC1=C2C(=CS1)C(=O)NC3=CC=CC=C3N2C(=O)CN4CCN(CC4)C.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 443.39
Percent Purity >98%
Chemical Name or Material Telenzepine dihydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.