Learn More
Selleck Chemical LLC Temozolomide 10mM/1mL 85622-93-1 CCRG81045, NSC 362856,TMZ
Supplier:  Selleck Chemical LLC S123710MM/1ML
Encompass_Preferred
Temozolomide (CCRG81045, NSC 362856, TMZ) is a monofunctional SN-1 alkylating agent that can modify nitrogen atoms in the DNA ring and the extracyclic oxygen group, chemically converted to MTIC and degrades to methyldiazonium cation, which transfers methyl groups to DNA at physiologic pH. A DNA damage inducer in L-1210 and L-1210/BCNU cells. Temozolomide induces apoptosis and exhibits antitumor activity.
Catalog No. 50-136-1056
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.