Learn More
Saf T Pak Inc Fortis 45-13.3L Refrigerated (2°-8°C) Insulated Shipping Solution
Supplier: Saf T Pak Inc 38474
FORTIS (2°-8°C, PCM6 and; Neopor)Designed for use when shipping high-risk, irreplaceable payloads, the FORTIS product line is our flagship 2°C to 8°C shipping system. Using a graphite-infused polystyrene-insulated chest, Neopor, and a unique Phase Change Material that phases between 2°C to 8°C, this robust system has been pre-qualified to ISTA 7D. This system can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900). By validating the shippers to the most challenging industry standard profiles, the Fortis is a cost effective solution that takes risk out of your supply chain.FEATURES: Tested to ISTA 7D Profiles 2°-8°C Universal Solution: 48 and 72 hour15°-25°C Solution: 48, 72, 96 hourAdvanced Phase Change MaterialPriced For Single Use - Robust Enough for Multiple ShipmentsTemp/Profile/Duration: 2-8/S72/W96
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.