Learn More
Saf T Pak Inc Sepio 5-1.72L Refrigerated (2-8C) Insulated Shipping Solution 4/CASE
Supplier: Saf T Pak Inc 24068
The cost-effective SEPIO is a tested system offering a low barrier of entry when shipping payloads needing refrigeration, and can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900).FEATURES: Key Features | ClaimsPre-qualified for 2C to 8C and 15C to 25C using the same brick cassettesInmark Legacy ProfilesSeasonal and; Universal pack-outsCorrugated Cassettes for Easy HandlingMultiple payload sizesCost-effective solutionsWater-based refrigerant cassettesGlobal availabilityTemp/Profile/Duration:2-8/S72/W72Outside Dimensions - Shipping Carton(mm) 314 x 314 x 332(in) 12.4 x 12.4 x 13.1Inside Dimensions - Product Space(mm) 177 x 127 x 76(in) 7.0 x 5.0 x 3.0Dead Weight (Includes Refrigerant)(kg) 3.8(lb) 8.4Winter (2-8C)Outside Dimensions - Shipping Carton(mm) 314 x 314 x 332(in) 12.4 x 12.4 x 13.1Inside Dimensions - Product Space(mm) 177 x 127 x 76(in) 7.0 x 5.0 x 3.0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.