Learn More
Chemglass Life Sciences Temperature Controller, Chemglass, PID, Type T, CONTROLLER ONLY
SDP
Supplier:  Chemglass Life Sciences CG1500110
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Temperature Controller, Chemglass, PID, Type T, CONTROLLER ONLY. Economical temperature controller for applications that do not require the precise temperature (± 0.1°). Auto tune PID process controller for use with Chemglass oil baths or mantles up to 5L. Controller has 1500 watts of power and will control temperatures to ± 1.0°C.
Catalog No. 50-401-785
missing translation for 'QuantityLeft'