Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco TERi Androgynous Geriatric Patient Simulator - Light
SDPSCI_ED

Catalog No. S97910 Shop All Simulaids, Inc. Products
Click to view available options
Color:
This item is not returnable. View return policy
This item is not returnable. View return policy

A comprehensive male/female elderly patient care trainer for medical simulation. Evolving from our GERi/KERi full body manikins, it has a more realistic look and feel accurately representing the human anatomy. Weight is distributed to represent a real patient for lifting and carrying.

This complete simulation comes with auscultation and patient monitoring.

Passive Range of Motion

  • Seating, laying, finger/toe manipulation with no effort from the patient

Cardiovascular Support

  • Carotid pulses, CPR with quality performance metrics and auscultation

Intravenous, Injection, Subcutaneous Injection Sites

  • Multiple locations equipped with standard arm IM injection pads

Nasogastric Tube Procedures

  • Placement
  • Lavage
  • Gavage
  • Feeding and suctioning

Enhanced Aesthetics

  • Soft silicone skin with noticeable normal and cancerous skin moles
  • Stage 1 sacral ulcer
  • Reddened skin folds

Respiratory Support

  • Manual ventilation with chest rise
  • Oxygen administration
  • Tracheostomy care
  • Airway/trach suctioning
  • Airway intubation, ventilation and auscultation.

Grooming and Daily Living Assistance

  • Bathing, clothing changes
  • Denture placement/removal
  • Hearing aid placement/removal
  • Eye irrigation and optic drops
  • Ostomy and urinary catheter care
  • Haircare

Indwelling Male and Female Urinary Catheterization

  • Includes support for urethral and suprapubic catheters

Advanced Skills

  • Urinary catheterization
  • Enema administration
  • Pelvic and prostate examination

Specifications

Product Type

Geriatric Patient Care Simulator

For Use With (Application)

Simulating Patient Care

Color

Light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.