Learn More
Enzo Life Sciences BIOARRAY™ Terminal labeling kit with biotin-ddUTP for DNA probe array assays (25Reactions)
Supplier: Enzo Life Sciences ENZ42630
The BIOARRAY™ 3’-OH Terminal Labeling System is the gold standard end-labeling system for use with Affymetrix DNA SNP, Resequencing and Prokaryotic microarrays. The BIOARRAY™ Terminal Labeling Kit with Biotin-ddUTP has been developed for DNA probe array assays. This method uses Bio-ddUTP and terminal deoxynucleotide transferase to catalyze the addition of a single biotin-ddUMP to the 3’-OH terminus of an amplified and fragmented target DNA molecule. A standard reaction will label up to 100 picomoles (equivalent to 1µg of a 30-nucleotide sequence). When DNA fragments are labeled at the 3’-OH terminus, the sequence bias that occurs with either nick translation or random priming is eliminated.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.