Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terpin Hydrate, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. p-7158366 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Pail

TE110, 2451-01-6, C10H20O2•H2O

Spectrum™ Chemical Terpin Hydrate, USP is used to control a cough associated with a cold or other respiratory illnesses. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2451-01-6
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
InChI Key JGKJMBOJWVAMIJ-UHFFFAOYSA-N
IUPAC Name 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol hydrate
SMILES O.CC(C)(O)C1CCC(C)(O)CC1

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 98 to 100.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.