Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-Methylcarbazate 97.0+%, TCI America™
SDP

Catalog No. p-7123293 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 21075-83-2
Molecular Formula C6H14N2O2
Molecular Weight (g/mol) 146.19
MDL Number MFCD05669700
InChI Key IHMQNZFRFVYNDS-UHFFFAOYSA-N
Synonym 1-tert-Butoxycarbonyl-1-methylhydrazine, 1-Boc-1-methylhydrazine, 2-Methylcarbazic Acid tert-Butyl Ester
PubChem CID 3748280
IUPAC Name tert-butyl N-amino-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)N

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material tert-Butyl 2-Methylcarbazate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.