Learn More
Hach Company Acidity Test Kit, Model AC-6
Supplier:  Hach Company 222301
Encompass_Preferred
Acidity is usually caused by the presence of mineral acids, salts of strong acids, or free carbon dioxide. In surface or ground waters, acidity may be form natural substances or industrial pollution. Excessive levels cause corrosion and can be detrimental to fish. This kit, which utilizes drop count titration, now reads results in mg/L, with a conversion factor to gpg.
Catalog No. 50-199-4728
missing translation for 'QuantityLeft'
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.