Learn More
Cole-Parmer Fluke 700PTP-1 Pneumatic Test Pump, -13 to 600 psi
Supplier:  Cole-Parmer 6845075
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
The Fluke 700PTP-1 pneumatic test pump is capable of supplying pressures up to 600 psi. For use with 700 series pressure modules or test gauges. Can be directly connected to the reference port consolidating two separate pieces when performing pressure calibration and testing. Can also be used with 717 pressure calibrators. The vernier can be used as a stand-alone low pressure calibration source. Available for sale in the United States and Canada only.
Catalog No. 50-993-407
missing translation for 'QuantityLeft'