Learn More
Eisco™ Test Strips - pH 0 to 2.5 - Ideal for Strong Acids
Eisco Labs pH 0 to 2.5 test strips provide precise measurement for strong acids. Ideal for quick and accurate testing.
Supplier: Eisco™ CH202403
Description
- Precision measurement: These test strips offer accurate measurement in 0.5 pH increments, allowing for precise monitoring of acidity levels
- Versatile usage: Specifically designed for testing strong acids, these strips are suitable for various industrial, laboratory, and educational purposes where acidity levels need to be determined
- Convenient packaging: Each bag contains 50 test strips, providing ample supply for multiple tests. The compact dimensions of the bag (3 x 4 in.) ensure easy storage and portability
- Reliable results: Precision label developed using NIST-traceable pH buffers at 25°C these colorimetric pH test strips are meticulously calibrated against pH-meter measurements for accuracy
- Shelf stable: With a minimum shelf-life of 3 years, these test strips maintain their effectiveness over an extended period, offering long-term usability and reliability in pH testing applications
- Our pH 0-2.5 Test Strips offer precise measurement in 0.5 pH increments, making them ideal for testing strong acids
- With 50 strips per bag, these strips provide convenience and ease of use
- Featuring a clear and pre-printed Precision label, these strips ensure accurate readings
- With a minimum shelf-life of 3 years, they offer long-term reliability
- Designed for buffered solutions, these strips are not suitable for non or low buffered solutions
- Each strip should be dipped into the solution for 1-2 seconds and compared immediately to the color chart provided
- Please note, these products are not approved for medical use and should not be used for diagnosing or treating medical conditions
Specifications
0 to 2.5 | |
pH 0-2.5 Test Strips |