Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium hydrogen sulfate, LiChropur™, MilliporeSigma™

Catalog No. M1183120025 Shop All MilliporeSigma Products
Click to view available options
Quantity:
Packaging:

Suitable for ion pair chromatography

Chemical Identifiers

CAS

32503-27-8

Molecular Formula

C16H37NO4S

Molecular Weight (g/mol)

339.54

InChI Key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Synonym

tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs

PubChem CID

94433

IUPAC Name

tetrabutylazanium hydrogen sulfate

SMILES

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point

168°C to 174°C

Color

Colorless

pH

1 to 2 (100g/L, H2O, 20°C)

Packaging

Glass Bottle Pre Packed

Quantity

25 g

Formula Weight

339.54

Percent Purity

>99.0%

Physical Form

Crystals

Chemical Name or Material

Tetra-n-butylammonium hydrogen sulfate

EINECSNumber

251-068-5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.