Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. p-7137829 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7182-86-7
Molecular Formula C23H43NO3S
Molecular Weight (g/mol) 413.661
MDL Number MFCD00043227
InChI Key REAVCZWUMGIGSW-UHFFFAOYSA-M
PubChem CID 10971700
IUPAC Name 4-methylbenzenesulfonate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CC1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 5 g
Formula Weight 413.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium p-Toluenesulfonate
EINECSNumber (2)-0186
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.