Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, ULTRA RESI-ANALYZED™, J.T. Baker™

Catalog No. 02004412 Shop All Avantor Products
Click to view available options
Quantity:
Packaging:

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Trace Hydrocarbon Analysis by IR Spectrophotometry; For Laboratory, Research, or Manufacturing Use

Chemical Identifiers

CAS

127-18-4

Molecular Formula

C2Cl4

Molecular Weight (g/mol)

165.82

MDL Number

MFCD00000834

InChI Key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Synonym

tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin

PubChem CID

31373

ChEBI

CHEBI:17300

IUPAC Name

1,1,2,2-tetrachloroethene

SMILES

ClC(Cl)=C(Cl)Cl

Specifications

Packaging

Narrow Mouth Amber Glass Bottle

Quantity

4L

Specific Gravity

1L = 1.62kg

Formula Weight

165.83

Grade

ULTRA RESI-ANALYZED™

Chemical Name or Material

Tetrachloroethylene

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.