Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoroisophthalic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7136742 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 1551-39-9
Molecular Formula C8F4O4
Molecular Weight (g/mol) 236.08
MDL Number MFCD00042379
InChI Key PGRIMKUYGUHAKH-UHFFFAOYSA-L
Synonym tetrafluoroisophthalic acid, 2,4,5,6-tetrafluoroisophthalic acid, tetrafluoroisophthalicacid, tetrafluorobenzene-1,3-dicarboxylic acid, tetrafluoroisophthalic, rarechem al bo 0259, acmc-1c1aw
PubChem CID 297552
IUPAC Name tetrafluorobenzene-1,3-dicarboxylate
SMILES [O-]C(=O)C1=C(F)C(C([O-])=O)=C(F)C(F)=C1F

Specifications

Melting Point 211°C
Color White
Quantity 1 g
Formula Weight 238.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoroisophthalic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.