Learn More
TARGETMOL CHEMICALS INC TETRAHYDROBERBERINE 50MG
Supplier:  TARGETMOL CHEMICALS INC T2874
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tetrahydroberberine (Canadine) is an isoquinoline alkaloid isolated from corydalis tuber has a micromolar affinity for dopamine D(2) receptors. purity: 99%
Catalog No. 50-257-7035