Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrahydrofuran, Low Water, HPLC Grade, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle with Septum
Description
- For use in liquid chromatography, organic and biosynthesis
Specifications
Specifications
Formula Weight | 72.11 |
Grade | HPLC |
Packaging | Narrow Mouth Amber Glass Bottle with Septum |
Specific Gravity | 1L = 0.88kg |
Flash Point | −14.00°C |
Quantity | 1L |
Chemical Name or Material | Tetrahydrofuran, Low Water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction