Learn More
Medchemexpress LLC HY-N0300 10mg Medchemexpress, Tetrahydropalmatine CAS:2934-97-6 Purity:>98%
Supplier: Medchemexpress LLC HYN030010MG
Medchemexpress, HY-N0300 10mg Tetrahydropalmatine CAS:2934-97-6 Tetrahydropalmatine, an active component isolated from corydalis, acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.