Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenyllead 97%, Thermo Scientific™

Catalog No. p-199136
Click to view available options
Quantity:
5 g
10 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 595-89-1
Molecular Formula C24H20Pb
Molecular Weight (g/mol) 515.60
MDL Number MFCD00002999
InChI Key WBJSMHDYLOJVKC-UHFFFAOYSA-N
Synonym tetraphenyllead, tetraphenyl lead, plumbane, tetraphenyl, pbph4, lead, tetraphenyl-,, lead tetraphenyl, acmc-209mel, wbjsmhdylojvkc-uhfffaoysa
PubChem CID 72906
ChEBI CHEBI:30184
IUPAC Name tetraphenylplumbane
SMILES C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 228°C to 231°C
Color White
Infrared Spectrum Authentic
Linear Formula Pb(C6H5)4
Quantity 5 g
Beilstein 16, 917
Formula Weight 515.6
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Tetraphenyllead, 97%
EINECSNumber 209-871-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.