Learn More
ACROBiosystems ACROBiosystems Human TFPI Protein, His Tag
Recombinant Protein;50UG;Human TFPI, His Tag (TFI-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Lys 282 (Accession # NP_006278.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.0 kDa. The protein migrates as 41-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.