Learn More
Gold Biotechnology Inc TG1 Phage Display Electrocompetent Cells 5x100 μl
SDP
Supplier:  Gold Biotechnology Inc CC2055X100C
Encompass_Preferred
GoldBio’s TG1 Phage Display Electrocompetent E. coli cells are suitable for protein expression and preparation of antibody or peptide phage display libraries. TG1 Phage Display cells have ≥1 x 109 cfu/µg efficiency with electroporation and have Amber suppressor strain (supE).
Catalog No. NC2942862
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.