Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF beta-1,2,3 Mouse anti-Human, Biotin, Clone: eBio16TFB, eBioscience™ Invitrogen™

Mouse Monoclonal Antibody
$117.00
Specifications
Antigen | TGF beta-1,2,3 |
---|---|
Clone | eBio16TFB |
Concentration | 0.5 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
13-992-381
![]() ![]() |
LIFE TECHNOLOGIES
13992381 |
50 μg |
Each of 1
for $117.00
Save 29% Reg : |
|
|||||
Description
TGF beta-1,2,3 Monoclonal antibody specifically detects TGF beta-1,2,3 in Human samples. It is validated for ELISASpecifications
TGF beta-1,2,3 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; tgf beta 1; tgf beta 3; tgf beta1; TGFB; Tgfb1; Tgfb-1; TGFB4; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; TGF-beta4; tissue growth factor-beta4; Transforming growth factor; transforming growth factor beta 1; transforming growth factor beta 4; transforming growth factor beta 4 precursor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta-1 precursor | |
TGFB1 | |
Primary | |
TGFB1 |
eBio16TFB | |
ELISA | |
Biotin | |
Mouse | |
Human | |
P01137 | |
7040 | |
IgG1 κ | |
Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title