Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ The Original™ Red Mechanics Gloves

Catalog No. p-7214847 Shop All Mechanix Wear Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Original™ gloves are built with carbon-infused synthetic leather for touchscreen capabilities, and breathable TrekDry™ to deliver a blend of dexterity and durability.

  • TrekDry™ is form-fitting to help keep hands cool and comfortable
  • Adjustable thermoplastic rubber (TPR) wrist closures create a secure fit
  • Reinforced thumb and index finger improve durability
  • Touchscreen capable synthetic leather for a combination of dexterity and durability
  • Ideal for maintenance repairs, equipement operation, and DIY home improvement
  • Machine washable

Specifications

Back Material TrekDry™
Cuff Material Thermoplastic Rubber (TPR)
Coating Material Uncoated
Color Red
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Small
For Use With (Application) Automotive Work, Maintenance Repairs, Equipment Operation, and DIY Home Improvements
Glove Cuff Style Hook and Loop
Series The Original™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.