Learn More
Thermo Scientific™ Hypersil GOLD™ C8 Javelin™ Guard Columns
Shop All Thermo Scientific Products- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
Features of Hypersil GOLD C8 Solid Phase
- Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
- Lower hydrophobicity
- Faster separations
- Excellent peak shape
- High efficiency
- Outstanding sensitivity
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Specifications
Particle Size
5 μm
Length (Metric)
10 mm
Diameter (Metric) Inner
4 mm
Stationary Phase
C8