Learn More
Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Enzyme Freezers
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.
Supplier: Thermo Scientific™ TSX2320EV
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Every enzyme freezer comes with a complete set of nine shelves with enzyme storage bins to optimize organization and retrieval of smaller materials.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 50% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Temperature range of -35° to -15°C, factory setpoint at -20°C
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
Specifications
Enzyme Freezer | |
650 L | |
Solid | |
9 | |
UL, cUL, CE | |
45 Enzyme Bins | |
Warranty varies by country, please contact us for details | |
28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm) | |
37.5 in. | |
95.3 cm | |
78.6 in. | |
199.6 cm | |
28 in. | |
71.1 cm | |
Wireless | |
Smart-Vue Compatible | |
CEE 7/7 | |
208-230V, 50Hz | |
Energy efficient | |
476 lb. | |
422 lb. |
23 cu. ft. | |
-25°C to -15°C (-20°C) | |
1 | |
9 | |
Microprocessor | |
TSX Series | |
Manual | |
37.5 x 28 x 78.6 in. (95.3 x 71.1 x 199.6 cm) | |
28.5 in. | |
72.3 cm | |
58 in. | |
147.3 cm | |
24 in. | |
61 cm | |
Optional | |
16 A | |
230 V | |
50 Hz | |
R290 | |
216 kg | |
191 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.