Learn More
Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Enzyme Freezers
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.
Supplier: Thermo Scientific™ TSX3020EV
Description
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers are designed with features that support sample protection and sustainability objectives for the storage of enzymes requiring -20°C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 50% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Temperature range of -35° to -15°C, factory setpoint at -20°C
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models
Specifications
TSX Series | |
29.2 cu. ft. | |
208-230V, 50Hz | |
50 Hz | |
CEE 7/7 | |
28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm) | |
220 kg | |
251 kg | |
R290 | |
Optional | |
28.5 in. | |
78.5 in. | |
199.4 cm | |
30 in. | |
Manual | |
86.4 cm | |
38.9 in. | |
99.0 cm | |
Wireless | |
9 |
UL, cUL, CE | |
827 L | |
230 V | |
15 A | |
38.9 x 34.0 x 78.5 in. (99.0 x 86.4 x 199.4 cm) | |
484 lb. | |
552 lb. | |
Microprocessor | |
Smart-Vue Compatible | |
-25°C to -15°C (-20°C) | |
72.3 cm | |
58.0 in. | |
147.3 cm | |
76.2 cm | |
34.0 in. | |
38.9 x 34.0 x 78.5 in. (99.0 x 86.4 x 199.4 cm) | |
High-Performance Freezer | |
Solid | |
1 | |
9 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.