Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Pre-drilled
For use with SPME applications to reduce septum coring
Supplier:  MilliporeSigma™ Supelco™ 23162U
 View more versions of this product
Catalog No. 11-101-0266
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
For use with:
  • Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
  • Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
  • Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
  • Influent sampling ports in gas chromatographs
Specifications
Specifications
3/8 in.
SPME
Pre-drilled
LB-2 Septa
9.5 mm
50/Pk.
Thermogreen™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC