Learn More
Thermo Scientific Thermo Scientific Thermolyne 5.8L C1 Benchtop Muffle Furnace
The Thermo Scientific Thermolyne Benchtop Muffle Furnace is ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating. It reaches a 1200°C maximum temperature and is available in two capacities for added flexibility. The built-in vent port removes contaminants and moisture to extend the life of the heating element and furnace.