Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thieno[2,3-b]thiophene 97.0+%, TCI America™
SDP

Catalog No. p-7137377 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 250-84-0
Molecular Formula C6H4S2
Molecular Weight (g/mol) 140.218
MDL Number MFCD02824126
InChI Key YHBTXTFFTYXOFV-UHFFFAOYSA-N
Synonym 1,6-Dithiapentalene
PubChem CID 136062
IUPAC Name thieno[2,3-b]thiophene
SMILES C1=CSC2=C1C=CS2

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 226°C
Quantity 1 g
Formula Weight 140.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Thieno[2,3-b]thiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.