Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thieno[3,2-b]thiophene-2-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7137644 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 40985-58-8
Molecular Formula C7H3NS2
Molecular Weight (g/mol) 165.228
MDL Number MFCD08059461
InChI Key UPRBPGCXCNBTQH-UHFFFAOYSA-N
Synonym 2-Cyanothieno[3,2-b]thiophene
PubChem CID 55252708
IUPAC Name thieno[3,2-b]thiophene-5-carbonitrile
SMILES C1=CSC2=C1SC(=C2)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 200 mg
Formula Weight 165.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.