Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimerosal, Thermo Scientific Chemicals

Catalog No. p-7042232
Click to view available options
Quantity:
10 g
25 g

Thimerosal, CAS # 54-64-8, is a sulfhydryl (thiol) oxidizing reagent with antibacterial and antifungal activities that is used to protect components from chemical change or microbial action.

Thimerosal, has been widely used as a preservative in a number of biological and drug products, including many vaccines, to help prevent potentially life threatening contamination with harmful microbes. It has been used as a preservative in vaccines, immunoglobulin preparations, skin test antigens, antivenins, ophthalmic and nasal products, and tattoo inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Thimerosal is an organic compound and a derivative of thiosalicyclic acid. It shows antibacterial and antifungal activities and has the ability to protect against chemical change or microbial action

• Thimerosal can inhibit the sulfhydryl-containing active site of some enzymes and bind to sulfhydryl compounds, such as glutathione, cysteine, and thiol groups of proteins

• It can activate the InsP3 calcium channel on the endoplasmic reticular membrane, triggering the release of calcium from intracellular stores, leading to a calcium-influx of extracellular calcium. Thus, thimerosal may stimulate or inhibit cellular functions dependent on calcium signaling

Application

• Thimerosal can increase histamine release and cell-mediated immunity

• It has the ability to inhibit various sulfhydryl-dependent enzymes and proteins, including PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na+/K+-dependent ATPases

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym thimerosal, thiomersal, thiomersalate, mercurothiolate, sodium merthiolate, sodium ethylmercurithiosalicylate, merthiolate, thimerosalate, thimerosalum, thimersalate
PubChem CID 16684434
ChEBI CHEBI:9546
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 234°C to 237°C
Color White
Flash Point 250°C (482°F)
Quantity 10 g
UN Number UN2025
Beilstein 8169555
Merck Index 14,9317
Solubility Information Soluble in water or ethanol
IUPAC Name sodium [(2-carboxylatophenyl)sulfanyl](ethyl)mercury
Formula Weight 404.81
Physical Form Powder
Chemical Name or Material Thimerosal
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H301-H310-H330-H373
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 200-210-4
RTECSNumber OV8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.