Learn More
Electron Microscopy Sciences Variable Size Geological Thin Section Holder, Hitachi M4
Supplier:  Electron Microscopy Sciences 7594116H
Encompass_Preferred
This thin section holder is made from vacuum grade aluminum with copper clips and brass screws . The unit measures 51x32x8 mm and holds either standard thin sections of 47 x 27 mm or smaller or larger thin sections. Smallest size would be 40x20 mm. The holder comes complete with 2 clips to insure nothing moves. The holder has convenient tweezer inserts which allow for easy loading.
Type: Hitachi M4
Catalog No. 50-190-1538
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.