Learn More
Thionicotinamide, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 185°C to 190°C |
Flash Point | 160°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 25 g |
Solubility Information | Solubility in water: 0.2g/100 mL (20°C) |
Formula Weight | 138.19 |
Percent Purity | 99% |
Physical Form | Powder |
Show More |
Safety and Handling
EINECSNumber | 225-036-6 |
RTECSNumber | QS4488000 |
TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.