Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thionyl chloride, 99%, Thermo Scientific Chemicals

Catalog No. p-4407981
Click to view available options
Quantity:
50 g
500 g
2 x 1 kg

Thionyl chloride is used as a chlorinating reagent for acids and alcohols. It is often used in the preparation of a wide variety of alkyl chloride and acyl chloride compounds, which are useful in the manufacture of fine chemicals, drugs, drug intermediates and vitamins. It is used to prepare thionyl bromide and thionyl fluoride. It finds use in several important organic transformations including pummerer rearrangement, and in lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7719-09-7
Molecular Formula Cl2OS
Molecular Weight (g/mol) 118.96
MDL Number MFCD00011449
InChI Key FYSNRJHAOHDILO-UHFFFAOYSA-N
Synonym thionyl chloride, sulfinyl chloride, sulfurous dichloride, thionylchloride, sulfurous oxychloride, sulfur oxychloride, sulfinyl dichloride, socl2, dichlorosulfoxide, sulfur chloride oxide
PubChem CID 24386
ChEBI CHEBI:29290
IUPAC Name thionyl dichloride
SMILES O=S(Cl)Cl

Specifications

Boiling Point 76°C
Melting Point °C to 104.5°C
Physical Form Liquid
Quantity 50 g
Assay Percent Range >99%
Solubility Information Miscible with toluene,chloroform,benzene,carbon tetrachloride and diethyl ether.
Formula Weight 118.97
Refractive Index 1.527
Sensitivity Moisture sensitive
Density 1.638 g/mL
Chemical Name or Material Thionyl chloride
Show More Show Less
Hazard Category H302-H314-H331-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 231-748-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.