Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thiophenol, Approx. 97%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
500 mL
50 kg
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Thiophenol , also known as Benzenethiol or Phenyl mercaptan, is an organosulfur compound and colorless liquid which is considered the simplest aromatic thiol. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS | 108-98-5 |
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Percent Purity | ∼97% |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction