Learn More
Bachem rec TNF-a (human)
SDP
Supplier:  Bachem 4031300.0050
Encompass_Preferred
rec TNF-a (human), 94948-59-1 , 50ug. This monocyte-macrophage-derived cytokine was first characterized by its in vivo antitumor properties and in vitro cytotoxicity to certain transformed cell lines. TNF-a also exerts a growth-enhancing effect on various cells and thus is a bifunctional growth regulator like e.g. IFN-a. As a mediator of inflammatory processes, TNF-a shares many similar biological activities with IL-1.
Catalog No. 50-194-6737
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.