Learn More
CUSABIO BIOTECHNOLOGY LLC Rat thyroxine,T4 ELISA Kit
Supplier:  CUSABIO BIOTECHNOLOGY LLC CSBE05082R
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
This Rat thyroxine (T4) ELISA Kit was designed for the quantitative measurement of Rat thyroxine (T4) protein in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. It is a Competitive ELISA kit, its detection range is 20 ng/mL-320 ng/mL and the sensitivity is 20 ng/mL. One kit contains 96 wells.
Catalog No. NC2952844