Learn More
Enzo Life Sciences Timosaponin AIII (5mg). CAS: 41059-79-4
SDP
Supplier:  Enzo Life Sciences BMLAP5040005
Encompass_Preferred
Induces autophagy in HeLa cells followed by apoptotic cell death (IC50=10μM). This cellular response has been found to be mediated via inhibition of mTORC1 and induction of ER stress (IC50 for BT474 cells=2.5µM and MDAMB231 cells=6µM). The pro-apoptotic response is selective for tumor cells over normal cells but autophagy is induced in both. Purity: ≥96% (HPLC). Solubility: Soluble in DMSO (20 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0320
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.